Benzaldehyde 2,4-Dinitrophenylhydrazone ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

B1593-1G B1593-10G
TCIAB1593-1GEA 23.1 EUR
TCIAB1593-1G TCIAB1593-10G
Benzaldehyde 2,4-Dinitrophenylhydrazone ≥98.0% (by titrimetric analysis)
Benzaldehyde 2,4-Dinitrophenylhydrazone
Formule: C₁₃H₁₀N₄O₄
MW: 286.25 g/mol
MDL Nummer: MFCD00156353
CAS nummer: 1157-84-2
UN: 1325
ADR: 4.1 III

Order Now

Specification Test Results

Appearance Light yellow to Brown powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 240.0 to 244.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR