(S)-5-Amino-2-((tert-butoxycarbonyl)amino)-5-oxopentanoic acid ≥98.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

B1649-10G B1649-25G
TCIAB1649-10GEA 47.3 EUR
TCIAB1649-10G TCIAB1649-25G
(S)-5-Amino-2-((tert-butoxycarbonyl)amino)-5-oxopentanoic acid ≥98.0% (by HPLC, titration analysis)
(S)-5-Amino-2-((tert-butoxycarbonyl)amino)-5-oxopentanoic acid
Formule: C₁₀H₁₈N₂O₅
MDL Nummer: MFCD00065571
CAS nummer: 13726-85-7
EINECS: 237-296-8

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Specific rotation [a]20/D -3.0 to -4.0 deg (C=2, EtOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR