Bis(4-methacryloylthiophenyl)sulphide ≥97.0% (by HPLC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: 4,4'-Thiodibenzenedithiol Dimethacrylate

B1662-25G B1662-5G
TCIAB1662-25GEA 1170 EUR
TCIAB1662-25G TCIAB1662-5G
Bis(4-methacryloylthiophenyl)sulphide ≥97.0% (by HPLC)
Bis(4-methacryloylthiophenyl)sulphide
Formule: C₂₀H₁₈O₂S₃
MW: 386.54 g/mol
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00142120
CAS nummer: 129283-82-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6fff77373902c37a88300fb31bd8ace52fb6e15d&vid=6fff77373902c37a88300fb31bd8ace52fb6e15d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 97.0 area%
Melting point 61.0 to 65.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR