Ethyl-2-(bromomethyl)acrylate ≥97.0% (by GC) stabilised
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: Ethyl 2-(bromomethyl)acrylate
Stabilisatie: Stabilised with hydroquinone (HQ)
B1820.0005 B1820-1G B1820-5G
TCIAB1820.0005EA 0 EUR
TCIAB1820.0005 TCIAB1820-1G TCIAB1820-5G
Ethyl-2-(bromomethyl)acrylate ≥97.0% (by GC) stabilised
Ethyl-2-(bromomethyl)acrylate
Formule: C₆H₉BrO₂
MW: 193.04 g/mol
Kookpunt: 86°C
Vlampunt: 81°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00031518
CAS nummer: 17435-72-2
Order Now
Specification Test Results
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %