ɑ,ɑ'-Bis(4-hydroxy-3,5-dimethylphenyl)-1,4-diisopropylbenzene ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

B1850-25G B1850-500G
TCIAB1850-25GEA 25.3 EUR
TCIAB1850-25G TCIAB1850-500G
ɑ,ɑ'-Bis(4-hydroxy-3,5-dimethylphenyl)-1,4-diisopropylbenzene ≥98.0% (by GC)
alpha,alpha'-Bis(4-hydroxy-3,5-dimethylphenyl)-1,4-diisopropylbenzene
Formule: C₂₈H₃₄O₂
MDL Nummer: MFCD01321133
CAS nummer: 36395-57-0

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 160.0 to 165.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR