4-tert-Butylstyrene ≥90.0% (by GC) stabilised

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing


Stabilisatie: Stabilised with 4-tert-butylpyrocatechol (TBC)

B1877-25ML B1877-500ML
TCIAB1877-25MLEA 23.1 EUR
TCIAB1877-25ML TCIAB1877-500ML
4-tert-Butylstyrene ≥90.0% (by GC) stabilised
4-tert-Butylstyrene
Formule: C₁₂H₁₆
Vlampunt: 81°C
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD00065126
CAS nummer: 1746-23-2
EINECS: 217-126-9

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 90.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR