3-Benzylthiazolium bromide ≥98.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

B1939-25G B1939-5G
TCIAB1939-25GEA 350 EUR
TCIAB1939-25G TCIAB1939-5G
3-Benzylthiazolium bromide ≥98.0% (by HPLC, titration analysis)
3-Benzylthiazolium bromide
Formule: C₁₀H₁₀BrNS
MDL Nummer: MFCD01631293
CAS nummer: 75066-50-1

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 155.0 to 159.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR