4,4'-Bis(ɑ,ɑ-dimethylbenzyl)diphenylamine ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: 4,4-Bis(α,α-dimethylbenzyl)diphenylamine

B1970-25G B1970-500G
TCIAB1970-25GEA 17.6 EUR
TCIAB1970-25G TCIAB1970-500G
4,4'-Bis(ɑ,ɑ-dimethylbenzyl)diphenylamine ≥98.0% (by GC)
4,4'-Bis(alpha,alpha-dimethylbenzyl)diphenylamine
Formule: C₃₀H₃₁N
MDL Nummer: MFCD00337918
CAS nummer: 10081-67-1
EINECS: 233-215-5

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 99.0 to 103.0 deg-C
Solubility in Acetonitrile almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR