(R)-1-Boc-2-Azetidinemethanol ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: (R)-1-(tert-Butoxycarbonyl)-2-azetidinemethanol, (R)-1-Boc-2-azetidinemethanol

B2174-1G B2174-100MG
TCIAB2174-1GEA 1250 EUR
TCIAB2174-1G TCIAB2174-100MG
(R)-1-Boc-2-Azetidinemethanol ≥98.0% (by GC)
(R)-1-Boc-2-Azetidinemethanol
Formule: C₉H₁₇NO₃
MDL Nummer: MFCD03790937
CAS nummer: 161511-90-6

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Optical purity (LC) min. 98.0 ee%
Specific rotation [a]20/D +72.0 to +78.0 deg (C=1, MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR