2,6-Bis((S)-4-phenyl-4,5-dihydrooxazol-2-yl)pyridine ≥95.0% (by HPLC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

B2220-1G B2220-250MG
TCIAB2220-1GEA 319 EUR
TCIAB2220-1G TCIAB2220-250MG
2,6-Bis((S)-4-phenyl-4,5-dihydrooxazol-2-yl)pyridine ≥95.0% (by HPLC)
2,6-Bis((S)-4-phenyl-4,5-dihydrooxazol-2-yl)pyridine
Formule: C₂₃H₁₉N₃O₂
MDL Nummer: MFCD01863586
CAS nummer: 174500-20-0

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 95.0 area%
Optical purity (LC) min. 98.0 ee%
Melting point 170.0 to 174.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR