3,5-Bis(3,5-dimethoxybenzyloxy)benzyl alcohol ≥94.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

B2246-1G B2246-5G
TCIAB2246-1GEA 80.3 EUR
TCIAB2246-1G TCIAB2246-5G
3,5-Bis(3,5-dimethoxybenzyloxy)benzyl alcohol ≥94.0% (by GC)
3,5-Bis(3,5-dimethoxybenzyloxy)benzyl alcohol
Formule: C₂₅H₂₈O₇
MDL Nummer: MFCD03844779
CAS nummer: 176650-92-3

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR