Bis(neopentyl glycolato)diboron ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

B2254-1G B2254-25G B2254-5G
TCIAB2254-1GEA 17.6 EUR
TCIAB2254-1G TCIAB2254-25G TCIAB2254-5G
Bis(neopentyl glycolato)diboron ≥98.0% (by GC)
Bis(neopentyl glycolato)diboron
Formule: C₁₀H₂₀B₂O₄
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD02093062
CAS nummer: 201733-56-4

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 186.0 to 193.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR