2-Benzyloxy-1,3-propanediol ≥97.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: 2-(Benzyloxy)propane-1,3-diol, 2-O-Benzylglycerol, Glycerol 2-Benzyl Ether

B2271-1G B2271-5G
TCIAB2271-1GEA 139 EUR
TCIAB2271-1G TCIAB2271-5G
2-Benzyloxy-1,3-propanediol ≥97.0% (by GC)
2-Benzyloxy-1,3-propanediol
Formule: C₁₀H₁₄O₃
MDL Nummer: MFCD00075493
CAS nummer: 14690-00-7

Order Now

Specification Test Results

Appearance White to Light yellow to Green powder to crystal
Purity (GC) min. 97.0 %
Melting point 38.0 to 42.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR