(1S,2S)-1,2-Bis(2,4,6-trimethylphenyl)ethylenediamine ≥97.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

B2317-100MG B2317-500MG
TCIAB2317-100MGEA 102 EUR
TCIAB2317-100MG TCIAB2317-500MG
(1S,2S)-1,2-Bis(2,4,6-trimethylphenyl)ethylenediamine ≥97.0% (by GC, titration analysis)
(1S,2S)-1,2-Bis(2,4,6-trimethylphenyl)ethylenediamine
Formule: C₂₀H₂₈N₂
MDL Nummer: MFCD06797064
CAS nummer: 186769-18-6

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 83.0 to 87.0 deg-C
Specific rotation [a]20/D -190 to -215 deg (C=1,CH2Cl2)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR