(+/-)-1,2-Bis(2,4,6-trimethylphenyl)ethylenediamine dihydrochloride ≥98.0% (by HPLC, total nitrogen)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: (+/-)-1,2-Dimesitylethylenediamine dihydrochloride, (+/-)-1,2-Diamino-1,2-dimesitylethane dihydrochloride

B2318-1G B2318-5G
TCIAB2318-1GEA 212 EUR
TCIAB2318-1G TCIAB2318-5G
(+/-)-1,2-Bis(2,4,6-trimethylphenyl)ethylenediamine dihydrochloride ≥98.0% (by HPLC, total nitrogen)
(+/-)-1,2-Bis(2,4,6-trimethylphenyl)ethylenediamine dihydrochloride
Formule: C₂₀H₂₈N₂.2HCl
MW: 369.37 g/mol
CAS nummer: 1820706-16-8

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR