Bis(2,2-diethoxyethyl)disulphide ≥90.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
B2806-1G B2806-5G
TCIAB2806-1GEA 74.8 EUR
TCIAB2806-1G TCIAB2806-5G
Bis(2,2-diethoxyethyl)disulphide ≥90.0% (by GC)
Bis(2,2-diethoxyethyl)disulphide
Formule: C₁₂H₂₆O₄S₂
Kookpunt: 155°C
Bewaar temperatuur: Vriezer
CAS nummer: 76505-71-0
Order Now
Specification Test Results
Appearance Light yellow to Yellow to Orange clear liquid
Purity (GC) min. 90.0 %