Glycerol dimethacrylate(mixture of 1,2- and 1,3-form) ≥90.0% (by GC) stabilised
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
Bis(methacryloyloxy)propanol
Stabilisatie:
Stabilised with 4-methoxyphenol (MEHQ)
B2938-25G
B2938-500G
TCIAB2938-25GEA
18.7
EUR
TCIAB2938-25G
TCIAB2938-500G
Glycerol dimethacrylate(mixture of 1,2- and 1,3-form) ≥90.0% (by GC) stabilised
Glycerol dimethacrylate(mixture of 1,2- and 1,3-form)
Formule:
C₁₁H₁₆O₅ Kookpunt: 104°C Vlampunt: 172°C |
MDL Nummer:
MFCD00066299 CAS nummer: 1830-78-0 EINECS: 217-388-4 |
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 90.0% (total of isomer) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...