2-Benzyloxybenzyl alcohol ≥97.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
B2943-25G B2943-5G
TCIAB2943-25GEA 328 EUR
TCIAB2943-25G TCIAB2943-5G
2-Benzyloxybenzyl alcohol ≥97.0% (by GC)
2-Benzyloxybenzyl alcohol
Formule: C₁₄H₁₄O₂
Kookpunt: 155°C
MDL Nummer: MFCD01664428
CAS nummer: 3381-87-1
Order Now
Specification Test Results
Appearance White or Colorless to Yellow powder to lump to clear liquid
Purity (GC) min. 97.0 %