6-Benzylamino-1-hexanol ≥97.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
B2968-25G
B2968-5G
TCIAB2968-25GEA
154
EUR
TCIAB2968-25G
TCIAB2968-5G
6-Benzylamino-1-hexanol ≥97.0% (by GC)
6-Benzylamino-1-hexanol
Formule:
C₁₃H₂₁NO |
CAS nummer:
133437-08-8 |
|
Specification Test Results
Purity (GC) | min. 97.0 % |
Appearance | Light yellow to Yellow to Orange clear liquid |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...