1-Benzyl-D-aspartate ≥98.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

B2993-1G B2993-5G
TCIAB2993-1GEA 35.2 EUR
TCIAB2993-1G TCIAB2993-5G
1-Benzyl-D-aspartate ≥98.0% (by HPLC, titration analysis)
1-Benzyl-D-aspartate
Formule: C₁₁H₁₃NO₄
Smeltpunt: 176°C
MDL Nummer: MFCD00037289
CAS nummer: 79337-40-9

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Optical purity (LC) min. 98.0 ee%
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR