H-Glu-otbu ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

B3000-25G B3000-5G
TCIAB3000-25GEA 389 EUR
TCIAB3000-25G TCIAB3000-5G
H-Glu-otbu ≥98.0% (by titrimetric analysis)
H-Glu-otbu
Formule: C₉H₁₇NO₄
MDL Nummer: MFCD00038562
CAS nummer: 45120-30-7

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 138.0 to 142.0 deg-C
Specific rotation [a]20/D +12.0 to +16.0 deg (C=1, water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR