N-Benzyl-2-naphthylamine ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
B3088-25G B3088-5G B3088-1G
TCIAB3088-25GEA 905 EUR
TCIAB3088-25G TCIAB3088-5G TCIAB3088-1G
N-Benzyl-2-naphthylamine ≥98.0% (by GC)
N-Benzyl-2-naphthylamine
Formule: C₁₇H₁₅N
Kookpunt: 190°C
MDL Nummer: MFCD00144848
CAS nummer: 13672-18-9
Order Now
Specification Test Results
Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 66.0 to 70.0 deg-C
Specificaties
Environmentally Preferable