4-Bromo-4',4''-dimethyltriphenylamine ≥95.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
4-Bromo-4,4-dimethyltriphenylamine, 4-Bromo-N,N-di-p-tolylaniline
B3089-1G
B3089-5G
TCIAB3089-1GEA
57.2
EUR
TCIAB3089-1G
TCIAB3089-5G
4-Bromo-4',4''-dimethyltriphenylamine ≥95.0% (by GC)
4-Bromo-4',4''-dimethyltriphenylamine
Formule:
C₂₀H₁₈BrN MW: 352.28 g/mol |
MDL Nummer:
MFCD03093258 CAS nummer: 58047-42-0 |
Specification Test Results
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 95.0 % |
Melting point | 100.0 to 105.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...