N4-Benzoylcytidine ≥98.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

B3094-1G B3094-5G
TCIAB3094-1GEA 34.1 EUR
TCIAB3094-1G TCIAB3094-5G
N4-Benzoylcytidine ≥98.0% (by HPLC, titration analysis)
N4-Benzoylcytidine
Formule: C₁₆H₁₇N₃O₆
Smeltpunt: 240°C
MDL Nummer: MFCD00010572
CAS nummer: 13089-48-0

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Specific rotation [a]20/D +43.0 to +48.0 deg (C=1, DMSO)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR