2-(Bromomethyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥90.0% (by GC, titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
(Bromomethyl)boronic Acid Pinacol Ester
B3199.0005
B3199-1G
B3199-5G
TCIAB3199.0005EA
0
EUR
TCIAB3199.0005
TCIAB3199-1G
TCIAB3199-5G
2-(Bromomethyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥90.0% (by GC, titration analysis)
2-(Bromomethyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Formule:
C₇H₁₄BBrO₂ MW: 220.90 g/mol Kookpunt: 68°C Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD09271779 CAS nummer: 166330-03-6 |
Specification Test Results
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 90.0 % |
Purity (Argentmetric Titration) | min. 90.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...