Bicalutamide ≥98.0% (by HPLC, total nitrogen)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
N-(4-Cyano-3-(trifluoromethyl)phenyl)-3-((4-fluorophenyl)sulphonyl)-2-hydroxy-2-methylpropanamide, N-[4-Cyano-3-(trifluoromethyl)phenyl]-3-[(4-fluorophenyl)sulphonyl]-2-hydroxy-2-methylpropionamide
B3206-200MG
B3206-1G
TCIAB3206-200MGEA
40.7
EUR
TCIAB3206-200MG
TCIAB3206-1G
Bicalutamide ≥98.0% (by HPLC, total nitrogen)
Bicalutamide
Formule:
C₁₈H₁₄F₄N₂O₄S MW: 430.37 g/mol |
MDL Nummer:
MFCD00869971 CAS nummer: 90357-06-5 Merck Index: 14,01200 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 194.0 to 198.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...