O-Benzoyl-N-methylhydroxylamine hydrochloride ≥98.0% (by HPLC, total nitrogen)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

B3239-1G B3239-5G
TCIAB3239-1GEA 53.9 EUR
TCIAB3239-1G TCIAB3239-5G
O-Benzoyl-N-methylhydroxylamine hydrochloride ≥98.0% (by HPLC, total nitrogen)
O-Benzoyl-N-methylhydroxylamine hydrochloride
Formule: C₈H₉NO₂
Smeltpunt: 136°C
CAS nummer: 27130-46-7

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR