5-tert-Butyl-1-pentafluorophenyl-N-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-glutamate ≥98.0% (by HPLC, total nitrogen)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: Fmoc-Glu(OtBu)-OPfp, Fmoc-Glu(OtBu)-Opfp, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-glutamic Acid 5-tert-Butyl 1-Pentafluorophenyl Ester, N-Fmoc-L-glutamic Acid 5-tert-Butyl 1-Pentafluorophenyl Ester, 5-tert-Butyl 1-Pentafluorophenyl N-Fmoc-L-glutamate, N-Fmoc-L-glutamic acid 5-tert-butyl ester pentafluorophenyl ester

B3318-1G B3318-5G
TCIAB3318-1GEA 40.7 EUR
TCIAB3318-1G TCIAB3318-5G
5-tert-Butyl-1-pentafluorophenyl-N-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-glutamate ≥98.0% (by HPLC, total nitrogen)
5-tert-Butyl-1-pentafluorophenyl-N-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-glutamate
Formule: C₃₀H₂₆F₅NO₆
MW: 591.53 g/mol
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00065647
CAS nummer: 86061-04-3

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 122.0 to 126.0 deg-C
Specific rotation [a]20/D -17.0 ~ -21.0 deg (C=1, CHCl3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR