N-(tert-Butoxycarbonyl)-D-proline succinimidyl ester ≥98.0% (by total nitrogen basis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: Boc-D-Pro-OSu, N-Boc-D-proline Succinimidyl Ester

B3414-1G B3414-5G
TCIAB3414-1GEA 27.5 EUR
TCIAB3414-1G TCIAB3414-5G
N-(tert-Butoxycarbonyl)-D-proline succinimidyl ester ≥98.0% (by total nitrogen basis)
N-(tert-Butoxycarbonyl)-D-proline succinimidyl ester
Formule: C₁₄H₂₀N₂O₆
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD00069687
CAS nummer: 102185-34-2

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 96.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Specific rotation [a]20/D +54.0 to +58.0 deg (C=1, Dioxane)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR