2,3-Bis(4-bromophenyl)-2-butenedinitrile ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
B3437-1G
B3437-5G
TCIAB3437-1GEA
79.2
EUR
TCIAB3437-1G
TCIAB3437-5G
2,3-Bis(4-bromophenyl)-2-butenedinitrile ≥98.0% (by GC)
2,3-Bis(4-bromophenyl)-2-butenedinitrile
Formule:
C₁₆H₈Br₂N₂ |
CAS nummer:
315203-26-0 UN: 3439 ADR: 6.1,III |
Specification Test Results
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 216.0 to 220.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...