2-Butyl-1-n-octanol ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: 2-Butyl-n-octyl Alcohol, 2-Butyloctan-1-ol

B3457-25ML B3457-500ML B3457-100ML
TCIAB3457-25MLEA 30.8 EUR
TCIAB3457-25ML TCIAB3457-500ML TCIAB3457-100ML
2-Butyl-1-n-octanol ≥98.0% (by GC)
2-Butyl-1-n-octanol
Formule: C₁₂H₂₆O
Kookpunt: 104°C
Smeltpunt: –80°C
Vlampunt: 110°C
MDL Nummer: MFCD00053508
CAS nummer: 3913-02-8
EINECS: 223-470-0
UN: 3082
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f8716730b7013d5dffdbd59d9e0619de0d6f8f0d&vid=f8716730b7013d5dffdbd59d9e0619de0d6f8f0d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR