3-Benzylamino-1-propanol ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

B3462-5G B3462-25G
TCIAB3462-5GEA 45.1 EUR
TCIAB3462-5G TCIAB3462-25G
3-Benzylamino-1-propanol ≥98.0% (by titrimetric analysis)
3-Benzylamino-1-propanol
Formule: C₁₀H₁₅NO
Kookpunt: 115°C
MDL Nummer: MFCD00020622
CAS nummer: 4720-29-0
EINECS: 225-210-1

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 95.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR