2-Bromo-4-fluorothiophene ≥95.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

B3680-200MG B3680-1G
TCIAB3680-200MGEA 235 EUR
TCIAB3680-200MG TCIAB3680-1G
2-Bromo-4-fluorothiophene ≥95.0% (by GC)
2-Bromo-4-fluorothiophene
Formule: C₄H₂BrFS
Kookpunt: 60°C
Bewaar temperatuur: Vriezer
CAS nummer: 32431-65-5
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR