2-Bromo-3-dodecylthiophene ≥97.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

B3691-5G B3691-1G
TCIAB3691-5GEA 59.4 EUR
TCIAB3691-5G TCIAB3691-1G
2-Bromo-3-dodecylthiophene ≥97.0% (by GC)
2-Bromo-3-dodecylthiophene
Formule: C₁₆H₂₇BrS
Kookpunt: 165°C
MDL Nummer: MFCD10000891
CAS nummer: 139100-06-4

Order Now

Specification Test Results

Appearance Colorless to Red to Green clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR