Bis(2-bromoethyl)ether ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: 1-Bromo-2-(2-bromoethoxy)ethane, 2-Bromoethyl ether
B3752-5G B3752-25G
TCIAB3752-5GEA 27.5 EUR
TCIAB3752-5G TCIAB3752-25G
Bis(2-bromoethyl)ether ≥98.0% (by GC)
Bis(2-bromoethyl)ether
Formule: C₄H₈Br₂O
MW: 231.93 g/mol
Kookpunt: 45°C
Vlampunt: 85°C
MDL Nummer: MFCD00039196
CAS nummer: 5414-19-7
EINECS: 226-504-2
Order Now
Specification Test Results
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %