Isopropyl palmitate tech. 85%, Technical Grade
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
Hexadecylic acid isopropyl ester, Palmitic acid isopropyl ester, Hexadecanoic acid isopropyl ester, Isopropyl Palmitate, Isopropyl hexadecanoate
L04276.AK
L04276.AU
L04276.0F
L04276.AKEA
27.6
EUR
L04276.AK
L04276.AU
L04276.0F
Isopropyl palmitate tech. 85%, Technical Grade
Isopropyl palmitate
Purity: 85%
CAS No.: 142-91-6
Molecular Formula: CH3(CH2)14CO2CH(CH3)2
Formula Weight: 298.51
Flash Point: 183°(361°F)
Refractive Index: 1.4380
Melting Point: 11-13°
Boiling Point: 160°/2mm
Beilstein Reference: 1786567
MDL No.: MFCD00008993
Formule:
C₁₉H₃₈O₂ Kookpunt: 160 °C (2 mmHg) Smeltpunt: 11…13 °C Dichtheid: 0,852 Vlampunt: 183 °C (361 °F) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00008993 CAS nummer: 142-91-6 EINECS: 205-571-1 |