1,3-Benzenedimethanethiol ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
B3854-1G B3854-5G
TCIAB3854-1GEA 58.3 EUR
TCIAB3854-1G TCIAB3854-5G
1,3-Benzenedimethanethiol ≥98.0% (by GC)
1,3-Benzenedimethanethiol
Formule: C₈H₁₀S₂
Kookpunt: 170°C
MDL Nummer: MFCD00012311
CAS nummer: 41563-69-3
Order Now
Specification Test Results
Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Iodometric Titration) min. 97.0 %