Bicyclo[4.2.0]octa-1,3,5-triene ≥94.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

B3863-1G B3863-200MG
TCIAB3863-1GEA 132 EUR
TCIAB3863-1G TCIAB3863-200MG
Bicyclo[4.2.0]octa-1,3,5-triene ≥94.0% (by GC)
Bicyclo[4.2.0]octa-1,3,5-triene
Formule: C₈H₈
Kookpunt: 150°C
Vlampunt: 36°C
MDL Nummer: MFCD01321219
CAS nummer: 694-87-1
UN: 3295
ADR: 3,III

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 94.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR