2-Bromo-1,3-bis(bromomethyl)benzene ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

B3913-25G B3913-5G
TCIAB3913-25GEA 641 EUR
TCIAB3913-25G TCIAB3913-5G
2-Bromo-1,3-bis(bromomethyl)benzene ≥98.0% (by GC)
2-Bromo-1,3-bis(bromomethyl)benzene
Formule: C₈H₇Br₃
MDL Nummer: MFCD00229246
CAS nummer: 25006-88-6
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 104.0 to 108.0 deg-C

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR