O-tert-Butyl-L-serine ≥97.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

B3917-5G B3917-1G
TCIAB3917-5GEA 149 EUR
TCIAB3917-5G TCIAB3917-1G
O-tert-Butyl-L-serine ≥97.0% (by HPLC, titration analysis)
O-tert-Butyl-L-serine
Formule: C₇H₁₅NO₃
Smeltpunt: 226°C
MDL Nummer: MFCD00066088
CAS nummer: 18822-58-7

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Nonaqueous Titration) min. 97.0 %
Specific rotation [a]20/D -15.0 to -18.0 deg (C=1, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR