Bupivacaine hydrochloride ≥98.0% (by HPLC, titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
B3925-5G
B3925-25G
TCIAB3925-5GEA
78.1
EUR
TCIAB3925-5G
TCIAB3925-25G
Bupivacaine hydrochloride ≥98.0% (by HPLC, titration analysis)
Bupivacaine hydrochloride
Formule:
C₁₈H₂₈N₂O·HCl Smeltpunt: 257°C |
MDL Nummer:
MFCD00078956 CAS nummer: 18010-40-7 EINECS: 241-917-8 UN: 2811 ADR: 6.1,II |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...