7-Bromoisatin ≥97.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
B3974-1G
B3974-5G
TCIAB3974-1GEA
17.6
EUR
TCIAB3974-1G
TCIAB3974-5G
7-Bromoisatin ≥97.0% (by GC)
7-Bromoisatin
Formule:
C₈H₄BrNO₂ Smeltpunt: 200°C |
MDL Nummer:
MFCD00774354 CAS nummer: 20780-74-9 |
Specification Test Results
Appearance | Orange to Amber to Dark red powder to crystal |
Purity (GC) | min. 97.0 % |
Purity (Neutralization titration) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...