4-Bromo-o-toluonitrile ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
B3993-5G
B3993-25G
TCIAB3993-5GEA
67.1
EUR
TCIAB3993-5G
TCIAB3993-25G
4-Bromo-o-toluonitrile ≥98.0% (by GC)
4-Bromo-o-toluonitrile
Formule:
C₈H₆BrN |
MDL Nummer:
MFCD00041461 CAS nummer: 67832-11-5 UN: 3439 ADR: 6.1,III |
Specification Test Results
Appearance | White to Light yellow powder to crystaline |
Purity (GC) | min. 98.0 % |
Melting point | 65.0 to 69.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...