8-Bromo-2'-deoxyguanosine ≥98.0% (by HPLC, total nitrogen)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
B4001-200MG
B4001-1G
TCIAB4001-200MGEA
49.5
EUR
TCIAB4001-200MG
TCIAB4001-1G
8-Bromo-2'-deoxyguanosine ≥98.0% (by HPLC, total nitrogen)
8-Bromo-2'-deoxyguanosine
Formule:
C₁₀H₁₂BrN₅O₄ Smeltpunt: 210°C Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD01630971 CAS nummer: 13389-03-2 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Specific rotation [a]20/D | -3.0 to -5.0 deg (C=1, 1mol/L NaOH sol.) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...