Boscalid ≥98.0% (by HPLC, total nitrogen)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: 2-Chloro-N-(4'-chlorobiphenyl-2-yl)nicotinamide

B4038-100MG B4038-1G
TCIAB4038-100MGEA 36.3 EUR
TCIAB4038-100MG TCIAB4038-1G
Boscalid ≥98.0% (by HPLC, total nitrogen)
Boscalid
Formule: C₁₈H₁₂Cl₂N₂O
MW: 343.21 g/mol
MDL Nummer: MFCD06795150
CAS nummer: 188425-85-6
UN: 3077
ADR: 9 III
Merck Index: 14,01352

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 147.0 to 151.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR