6-Bromonicotinonitrile ≥97.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

B4039-1G B4039-5G
TCIAB4039-1GEA 24.2 EUR
TCIAB4039-1G TCIAB4039-5G
6-Bromonicotinonitrile ≥97.0% (by GC)
6-Bromonicotinonitrile
Formule: C₆H₃BrN₂
MDL Nummer: MFCD00234141
CAS nummer: 139585-70-9
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 97.0 %
Melting point 143.0 to 147.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR