5-Bromo-3-nitro-2-picoline ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: 5-Bromo-2-methyl-3-nitropyridine

B4070-1G B4070-5G
TCIAB4070-1GEA 27.5 EUR
TCIAB4070-1G TCIAB4070-5G
5-Bromo-3-nitro-2-picoline ≥98.0% (by GC)
5-Bromo-3-nitro-2-picoline
Formule: C₆H₅BrN₂O₂
MDL Nummer: MFCD09031419
CAS nummer: 911434-05-4

Order Now

Specification Test Results

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (GC) min. 98.0 %
Melting point 38.0 to 42.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR