1-Butyl-3-methylimidazolium thiocyanate ≥97.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

B4091-5G B4091-25G
TCIAB4091-5GEA 28.6 EUR
TCIAB4091-5G TCIAB4091-25G
1-Butyl-3-methylimidazolium thiocyanate ≥97.0% (by titrimetric analysis)
1-Butyl-3-methylimidazolium thiocyanate
Formule: C₉H₁₅N₃S
MDL Nummer: MFCD06798181
CAS nummer: 344790-87-0

Order Now

Specification Test Results

Appearance Light yellow to Yellow to Orange clear liquid
Purity (HPLC) min. 96.0 area%
Purity (Nonaqueous Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR