4-Bromo-2,6-diisopropylaniline ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

B4113-1G B4113-5G
TCIAB4113-1GEA 36.3 EUR
TCIAB4113-1G TCIAB4113-5G
4-Bromo-2,6-diisopropylaniline ≥98.0% (by titrimetric analysis)
4-Bromo-2,6-diisopropylaniline
Formule: C₁₂H₁₈BrN
Kookpunt: 110°C
MDL Nummer: MFCD09743895
CAS nummer: 80058-84-0

Order Now

Specification Test Results

Appearance Light yellow to Brown clear liquid
Purity (GC) min. 95.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR