1,2-Benzenedisulphonic imide ≥98.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

B4139-1G B4139-5G
TCIAB4139-1GEA 248 EUR
TCIAB4139-1G TCIAB4139-5G
1,2-Benzenedisulphonic imide ≥98.0% (by HPLC, titration analysis)
1,2-Benzenedisulphonic imide
Formule: C₆H₅NO₄S₂
MDL Nummer: MFCD07779379
CAS nummer: 4482-01-3

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 194.0 to 198.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR